missing translation for 'onlineSavingsMsg'
Learn More

CD61 (Integrin beta 3), PE, clone: 2C9.G3, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0611-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15268569

  • £68.21 / 50µg


EU Stock 7
Add to basket

Description

Description

Description: The 2C9.G3 (HMb3-1) monoclonal antibody reacts with mouse and rat CD61, also known as the integrin beta3. CD61 is expressed by activated T cells, granulocytes, and platelet. CD61 associates non-covalently with two integrin alpha subunits; alphaV (CD51) to form Vitronectin Receptor and alphaIib (CD41) to form gpIIb/IIIa. These heterodimeric complexes are responsible for adhesion to extracellular matrix components including fibrinogen, fibronectin, fibronectin, vitronectin, thrombospondin and von Willebrand factor.Applications Reported: The 2C9.G3 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 2C9.G3 antibody has been tested by flow cytometric analysis of mouse splenocytes and bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD61 (Integrin beta 3)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O54890
BDPLT2, CD61, GP3A, GPIIIa, GT, integrin beta-3, platelet glycoprotein IIIa, platelet membrane glycoprotein IIIa, ITGB3
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
MurineþRat
Flow Cytometry
2C9.G3
PE
Itgb3
Conjugated
Itgb3
IgG
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
16416
Videos

For Research Use Only.