missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD61 Armenian Hamster anti-Mouse, Oligo, Clone: 2C9.G2 (also known as HMβ3-1), BD™ AbSeq ™
Armenian Hamster Monoclonal Antibody
Brand: BD Biosciences 940193
This item is not returnable.
View return policy
Description
CD61 Monoclonal antibody specifically detects CD61 in Mouse samples. It is validated for Single Cell 3' Sequencing
Specifications
| CD61 | |
| Monoclonal | |
| Oligo | |
| Itgb3; Integrin beta-3; INGRB3; Platelet glycoprotein IIIa; GP3A; GPIIIa | |
| Mouse T-cell Hybridoma 2B4 Vitronectin Receptor | |
| RUO | |
| Mouse | |
| IgG1 κ |
| Single Cell 3' Sequencing | |
| Clone 2C9.G2 (also known as HMβ3-1) | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Armenian Hamster | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction