missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD6 Rat anti-Mouse, BV605, Clone: J90-462, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 747538
This item is not returnable.
View return policy
Description
CD6 Monoclonal antibody specifically detects CD6 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| J90-462 | |
| Brilliant Violet 605 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction