missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD6 Rabbit anti-Human, Clone: C6/2884R, Novus Biologicals™
Rabbit Monoclonal Antibody
£403.00
Specifications
| Antigen | CD6 |
|---|---|
| Clone | C6/2884R |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
CD6 Monoclonal specifically detects CD6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CD6 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research, Immunology, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 923 | |
| IgG | |
| Protein A or G purified |
| C6/2884R | |
| Flow Cytometry 1 to 2 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
| Human recombinant full-length CD6 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title