missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD6 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5149359

Product Code. 30535265

  • £315.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

CD6, also known as T12, is a member of the scavenger receptor superfamily found on T and B cell subsets, thymocytes, and acute lymphocytic leukemia cells (ALL). CD6 interacts with its ligand CD166/ALCAM (activated leukocyte cell adhesion molecule) and serves as a coreceptor for T cell activation and stabilizer of the immunological synapse. CD6-ALCAM mediated cell adhesion is also important for T cell proliferation. CD6 may exert some its functions via association with CD5, probably by fine-tuning CD5 signaling. Ligation of CD6 has antiapoptotic role in chronic lymphocytic leukemia B cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD6
Polyclonal
Unconjugated
CD6
Cd6; CD6 antigen; CD6 antigen, Tp120; Cd6 molecule; FLJ44171; OX52; Soluble CD6; T12; T-cell differentiation antigen CD6; TP120
Goat
Antigen affinity chromatography
RUO
12511
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Inhibition Assays, Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q61003
CD6
Mouse myeloma cell line NS0-derived recombinant mouse CD6, Leu18-Gly396.
100 μg
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.