missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD6 Mouse anti-Rat, BV480, Clone: OX-52, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 746670
This item is not returnable.
View return policy
Description
CD6 Monoclonal antibody specifically detects CD6 in Rat samples. It is validated for Flow Cytometry.
Specifications
| CD6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Rat | |
| Purified |
| Flow Cytometry | |
| OX-52 | |
| Brilliant Violet 480 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido