missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD6 Monoclonal Antibody (MEM-98), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528095
This item is not returnable.
View return policy
Description
Mouse anti Human CD6 antibody, clone MEM-98 recognizes human CD6, also known as T12 or TP120.
CD6, also known as T12, is a member of the scavenger receptor superfamily found on T and B cell subsets, thymocytes, and acute lymphocytic leukemia cells (ALL). CD6 interacts with its ligand CD166/ALCAM (activated leukocyte cell adhesion molecule) and serves as a coreceptor for T cell activation and stabilizer of the immunological synapse. CD6-ALCAM mediated cell adhesion is also important for T cell proliferation. CD6 may exert some its functions via association with CD5, probably by fine-tuning CD5 signaling. Ligation of CD6 has antiapoptotic role in chronic lymphocytic leukemia B cells.
Specifications
| CD6 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 1% BSA and 0.09% sodium azide | |
| P30203 | |
| CD6 | |
| Purified CD6 antigen. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| MEM-98 | |
| FITC | |
| CD6 | |
| Cd6; CD6 antigen; CD6 antigen, Tp120; Cd6 molecule; FLJ44171; OX52; Soluble CD6; T12; T-cell differentiation antigen CD6; TP120 | |
| Mouse | |
| Protein A | |
| RUO | |
| 923 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction