missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD6 Antibody (rC6/372), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20439-20ug
This item is not returnable.
View return policy
Description
CD6 Monoclonal antibody specifically detects CD6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD6 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
| Recombinant full-length human CD6 protein | |
| 20 μg | |
| Cytokine Research, Immunology, Signal Transduction | |
| 923 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rC6/372 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction