missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD6 Antibody (96123), Janelia Fluor™ 549, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB727I
This item is not returnable.
View return policy
Description
CD6 Monoclonal antibody specifically detects CD6 in Mouse samples. It is validated for Western Blot, Flow Cytometry
Specifications
| CD6 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD6, Leu18-Gly396, Accession # Q61003 | |
| 0.5 mL | |
| Cytokine Research, Immunology, Signal Transduction | |
| 923 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry | |
| 96123 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction