missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5L, Mouse anti-Human, Clone: CD5L/2991, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79707-100UG
This item is not returnable.
View return policy
Description
CD5L Monoclonal specifically detects CD5L in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| CD5L | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| O43866, O43866 | |
| CD5L | |
| Recombinant fragment of human CD5L protein (around aa 269-366) (exact sequence is proprietary) | |
| 100 μg | |
| Apoptosis | |
| 922 | |
| Store at 4°C. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CD5L/2991 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| AIM, API6, apoptosis inhibitor 6, CD5 antigen-like, CD5 antigen-like (scavenger receptor cysteine rich family), CD5 molecule-like, CT-2, IgM-associated peptide, PRO229, Spalpha, SP-ALPHA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction