missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD5L Antibody Pair [HRP]
Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Description
This solid phase sandwich ELISA utilizes an antibody specific for Mouse CD5L.
Specifications
Specifications
| Assay Range | 6.25 pg/mL |
| Assay Sensitivity | 6.25-400 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes an antibody specific for Mouse CD5L. |
| Target Species | Mouse |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | AIM, API6, apoptosis inhibitor 6, CD5 antigen-like, CD5 antigen-like (scavenger receptor cysteine rich family), CD5 molecule-like, CT-2, IgM-associated peptide, PRO229, Spalpha, SP-ALPHA |
| Gene ID (Entrez) | 922 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction