missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5L Antibody (CD5L/2991), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79841APC
This item is not returnable.
View return policy
Description
CD5L Monoclonal specifically detects CD5L in Human samples. It is validated for Flow Cytometry.
Specifications
| CD5L | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| CD5L | |
| Recombinant fragment of human CD5L protein (around aa 269-366) (exact sequence is proprietary) (Uniprot: O43866) | |
| 0.1 mL | |
| Apoptosis | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| CD5L/2991 | |
| Flow Cytometry | |
| AIM, API6, apoptosis inhibitor 6, CD5 antigen-like, CD5 antigen-like (scavenger receptor cysteine rich family), CD5 molecule-like, CT-2, IgM-associated peptide, PRO229, Spalpha, SP-ALPHA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction