missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5L Antibody (38), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90356CL1
This item is not returnable.
View return policy
Description
CD5L Monoclonal antibody specifically detects CD5L in Mouse samples. It is validated for ELISA
Specifications
| CD5L | |
| Monoclonal | |
| CoraFluor 1 | |
| AIM, API6, apoptosis inhibitor 6, CD5 antigen-like, CD5 antigen-like (scavenger receptor cysteine rich family), CD5 molecule-like, CT-2, IgM-associated peptide, PRO229, Spalpha, SP-ALPHA | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD5L (Met 1-Val 352; Accession#: NP_033820.2) | |
| 0.1 mL | |
| Apoptosis | |
| 922 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 38 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction