missing translation for 'onlineSavingsMsg'
Learn More

CD5L Antibody (38), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18373940 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373940 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373940 Supplier Novus Biologicals Supplier No. NBP290356AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD5L Monoclonal specifically detects CD5L in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD5L
Applications ELISA
Classification Monoclonal
Clone 38
Conjugate Alexa Fluor 488
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias AIM, API6, apoptosis inhibitor 6, CD5 antigen-like, CD5 antigen-like (scavenger receptor cysteine rich family), CD5 molecule-like, CT-2, IgM-associated peptide, PRO229, Spalpha, SP-ALPHA
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD5L (Met 1-Val 352; NP_033820.2) .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 922
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.