missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD59 (Protectin) Monoclonal Antibody (OV9A2), Super Bright™ 645, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 64-0596-41
This item is not returnable.
View return policy
Description
Mouse Monoclonal Antibody
CD59 (Protectin) Monoclonal antibody specifically detects CD59 (Protectin) in Human samples. It is validated for Flow Cytometry
Specifications
| CD59 (Protectin) | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P13987 | |
| CD59 | |
| Affinity chromatography | |
| RUO | |
| 966 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| OV9A2 | |
| Super Bright 645 | |
| CD59 | |
| 16.3A5; 1F5; 1F5 antigen; 20 kDa homologous restriction factor; AA987121; Cd59; CD59 glycoprotein; Cd59a; CD59A glycoprotein; Cd59b; CD59B glycoprotein; EJ16; EJ30; EL32; G344; HRF20; HRF-20; MACIF; MAC-inhibitory protein; MACIP; MAC-IP; MEM43; MEM43 antigen; Membrane attack complex inhibition factor; Membrane inhibitor of reactive lysis; MIC11; MIN1; MIN2; MIN3; MIRL; MSK21; p18-20; protectin | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction