missing translation for 'onlineSavingsMsg'
Learn More

CD59 Mouse anti-Human, PE, Clone: P282 (H19), BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 555764

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 15898418

  • £349.00 / Each
Estimated Shipment: 19-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The p282 (HI9) monoclonal antibody specifically binds to CD59, a 19 kDa glycosylphosphatidylinositol (GPI)-anchored glycoprotein, expressed on hematopoietic and non-hematopoietic cells. Because of its role in interaction with complement activated products, CD59 has been termed membrane-attack-complex-inhibitory factor (MACIF), homologus restriction factor (HRF20), membrane inhibitor of reactive lysis (MIRL) and Protectin. It inhibits the cytolytic activity of complement by binding to C8 and C9, thereby blocking the assembly of the membrane attack complex. CD59 also participates in spontaneous T-cell/erythrocyte adhesion, interacts with CD2 and plays a role in T-cell activation.

Specifications

Specifications

CD59
Monoclonal
PE
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
P282 (H19)
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human Erythrocytes
100 Tests
Immunology
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.