missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD58 Recombinant Rabbit Monoclonal Antibody (23GB3050), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CD58 |
|---|---|
| Clone | 23GB3050 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089315
![]() |
Invitrogen™
MA553314 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089945
|
Invitrogen™
MA553315 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD58 Recombinant Monoclonal antibody specifically detects CD58 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| CD58 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P19256 | |
| 965 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3050 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Ag3; CD305; CD58; CD58 antigen; CD58 antigen, (lymphocyte function-associated antigen 3); CD58 molecule; FLJ23181; FLJ43722; LAIR1; LAIR-1; LFA3; LFA-3; Lymphocyte function-associated antigen 3; membrane receptor; RP5-1086K13.2; surface glycoprotein LFA-3 | |
| A synthesized peptide derived from CD58 (51-82AA). | |
| Primary | |
| -20°C | |
| CD58 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title