missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 Antibody (NK1/7565), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20869PCP
This item is not returnable.
View return policy
Description
CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD57 | |
| Monoclonal | |
| PerCP | |
| HNK1, HNK-1, Human Natural Killer-1, L2, LEU7, LEU-7 | |
| Recombinant fragment (around aa1-200) of human CD57 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Cell Biology, Immunology | |
| 27087 | |
| Store at 4°C in the dark. | |
| IgG2 λ |
| Immunohistochemistry (Paraffin) | |
| NK1/7565 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction