missing translation for 'onlineSavingsMsg'
Learn More

CD57 Antibody (NK1/7565), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18546099 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18546099 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18546099 Supplier Novus Biologicals Supplier No. NBP320869JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD57
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NK1/7565
Conjugate Janelia Fluor 646
Formulation 50 mM Sodium Borate
Gene Alias HNK1, HNK-1, Human Natural Killer-1, L2, LEU7, LEU-7
Host Species Mouse
Immunogen Recombinant fragment (around aa1-200) of human CD57 protein (exact sequence is proprietary)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 27087
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.