missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 Antibody (HNK-1 or Leu-7) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48346
This item is not returnable.
View return policy
Description
CD57 Monoclonal antibody specifically detects CD57 in Human, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
| CD57 | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| B3GAT1 | |
| Human peripheral blood mononuclear cells | |
| 7 mL | |
| Cancer, Cell Biology, Immunology | |
| 27087 | |
| Human, Rat (Negative) | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| HNK-1 or Leu-7 | |
| Immunohistochemistry-Paraffin | |
| HNK1, HNK-1, Human Natural Killer-1, L2, LEU7, LEU-7 | |
| Mouse | |
| Protein L or PEG purified | |
| RUO | |
| Primary | |
| Anti-CD57 marks a subset of lymphocytes known as natural killer (NK) cells. Follicular center cell lymphomas often contain many NK cells within the neoplastic follicles. Anti-CD57 also stains neuroendocrine cells and their derived tumors, including carcinoid tumor and medulloblastoma. Anti-CD57 can also be useful in separating type B3 thymoma from thymic carcinoma when combined with a panel that includes antibodies against GLUT1, CD5, and CEA. | |
| Store at 4C. | |
| IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction