missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 Antibody (HNK-1 or Leu-7) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
CD57 Monoclonal specifically detects CD57 in Human, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
Specifications
| Antigen | beta-1,3-Glucuronyltransferase 1/B3GAT1 |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Clone | HNK-1 or Leu-7 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin : 2 - 4 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml |
| Formulation | PBS with No Preservative |
| Gene Alias | Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase |
| Gene Symbols | B3GAT1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?