missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD56 (NCAM) Recombinant Mouse Monoclonal Antibody (CMSSB), Invitrogen™

Mouse Recombinant Monoclonal Antibody

£97.75 - £270.00

Specifications

Antigen CD56 (NCAM)
Clone CMSSB
Concentration 1.0 mg/mL
Applications Flow Cytometry, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30131837
View Documents
Invitrogen™
740024M
100 μL
£270.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30131661
View Documents
Invitrogen™
740024M20UG
20 μL
£97.75
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. Applications Tested: This CMSSB clone has been tested for immunocytochemistry at 10 μg/mL and for flow cytometry at 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

CD56 (NCAM) Recombinant Monoclonal antibody specifically detects CD56 (NCAM) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications

Specifications

CD56 (NCAM)
1.0 mg/mL
Recombinant Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.4
adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM
IgG1 κ
Protein A/G
Antibody
CMSSB
Flow Cytometry, Immunocytochemistry
Unconjugated
Mouse
Human
P13591
4684
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Ncam1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.