missing translation for 'onlineSavingsMsg'
Learn More

CD56 (NCAM), PE, clone: CMSSB, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0567-42

Code : Z2

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15311916

  • £159.14 / 100 tests


EU Stock 17
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This CMSSB monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells. Staining with CMSSB does not block binding of MEM188 or CB56.Applications Reported: This CMSSB antibody has been reported for use in flow cytometric analysis.Applications Tested: This CMSSB antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD56
CMSSB
PE
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Neural cell adhesion molecule
IgG1
RUO
Primary
Human
Videos