Learn More
CD56 (NCAM-1) Mouse, anti-Human, R718, Clone: B159, BD Horizon™
Mouse Monoclonal Antibody
£112.05 - £185.43
Specifications
| Antigen | CD56 (NCAM-1) |
|---|---|
| Clone | B159 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | R718 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17257754
|
BD Biosciences
566966 |
25 Tests |
£117.00 £112.05 / 25 tests Save £4.95 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17247754
|
BD Biosciences
566965 |
100 Tests |
£1225.00 £185.43 / 100 tests Save £1039.57 84% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.
Specifications
| CD56 (NCAM-1) | |
| Flow Cytometry | |
| R718 | |
| Mouse | |
| Human | |
| Refrigerate Upon Arrival |
| B159 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.