missing translation for 'onlineSavingsMsg'
Learn More

CD56 Mouse anti-Human, PE, Clone: B159, BD

Mouse Monoclonal Antibody

£88.51 - £247.89

Specifications

Antigen CD56
Clone B159
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15881449
View Documents
BD Biosciences
561903
25 Tests
ONLINE EXCLUSIVE PRICE
£92.05
£88.51 / Each
Save £3.54
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15818238
View Documents
BD Biosciences
555516
100 Tests
ONLINE EXCLUSIVE PRICE
£256.00
£247.89 / Each
Save £8.11
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.

Specifications

Specifications

CD56
Flow Cytometry
PE
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39
B159
Monoclonal
Mouse
Human
Human NK Cells
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.