Learn More
CD55, PE, clone: 67 (N-L063), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD55044
Code : 50
Additional Details : Weight : 0.50000kg
Description
CD55 (decay-accelerating factor, DAF) is a GPI-anchored membrane glycoprotein that protects autologous cells from classical and alternative pathway of complement cascade. Bidirectional interactions between CD55 and CD97 are involved in T cell regulation and CD55 can still regulate complement when bound to CD97. In tumours, besides protection agains complement, CD55 promotes neoangiogenesis, tumorigenesis, invasiveness and evasion of apoptosis.
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.Specifications
CD55 | |
Monoclonal | |
PE | |
CD55 | |
RP11-357P18.1, CR, CROM, DAF, TC | |
Mouse | |
Purified | |
ASR, ASR/IVD | |
1604 | |
Antibody | |
IgG1 |
Flow Cytometry | |
67 (N-L063) | |
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide | |
P08174 | |
CD55 | |
Human CD55 | |
2 mL | |
Primary | |
Human | |
Liquid |