missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD55 Monoclonal Antibody (MEM-118), Alexa Fluor™ 488
GREENER_CHOICE

Product Code. 15337414
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15337414 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15337414 Supplier Invitrogen™ Supplier No. MA518146

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes an epitope in SCR4 domain of CD55 (Decay accelerating factor, DAF), a 60-70 kDa glycosylphosphatidylinositol (GPI)-anchored single chain extracellular glycoprotein. CD55 is widely expressed on hematopoietic and on many non-hematopoietic cells; it is weakly present on NK cells.

CD55 (decay-accelerating factor, DAF) is a GPI-anchored membrane glycoprotein that protects autologous cells from classical and alternative pathway of complement cascade. Bidirectional interactions between CD55 and CD97 are involved in T cell regulation and CD55 can regulate complement when bound to CD97. In tumors, CD55 promotes neoangiogenesis, tumorigenesis, invasiveness and evasion of apoptosis. CD55 is a 70 kDa glycoprotein (in erythrocytes) anchored in the membrane by glycosylphosphatidylinositol tail. In other cells, the apparent molecular weight is somewhat larger for CD55 and has a substantial content of O-glycans. CD55 binds to activated C4b or C3b complement fragments on the cell surface, preventing the assembly and accelerating the decay of both classical and alternative pathways. CD55 carries the Cromer related blood group antigens. Two alternatively spliced transcripts encoding CD55 have been identified, and the predominant transcript encodes a membrane-bound protein expressed on cells exposed to plasma component proteins but an alternatively spliced transcript produces a soluble protein present at much lower levels. CD55 has been studied to have a role in prostate cancer growth and survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD55
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-118
Concentration 0.24 mg/mL
Conjugate Alexa Fluor 488
Formulation TBS with 0.2% BSA and 15mM sodium azide
Gene CD55
Gene Accession No. P08174
Gene Alias CD55; CD55 antigen; CD55 molecule (Cromer blood group); CD55 molecule, decay accelerating factor for complement; CD55 molecule, decay accelerating factor for complement (Cromer blood group); Cd55a; Complement decay-accelerating factor; complement decay-accelerating factor, GPI-anchored; complement-glycosylphosphatidylinositol; CR; CROM; Cromer blood group; Daf; Daf1; DAF-GPI; decay accelarating factor 1; decay accelerating factor 1; decay accelerating factor GPI-form; decay-accelarating factor; glycosylphophatidylinositol-anchored form; GPI anchor addition signal; GPI-DAF; RP11-357P18.1; TC
Gene Symbols CD55
Host Species Mouse
Immunogen HPB-ALL human T cell line.
Purification Method Size-exclusion chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1604
Target Species Human, Monkey
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.