missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD54 (ICAM-1) Monoclonal Antibody (1A29), PerCP-eFluor™ 710, eBioscience™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ 46-0540-80

Product Code. 15311270

  • £105.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Description: The 1A29 monoclonal antibody reacts with rat CD54, which is also known as intercellular adhesion molecule 1 (ICAM-1). In the immune system, this type I immunoglobulin (Ig)-like transmembrane protein is expressed on lymphocytes, macrophages, dendritic cells, monocytes, granulocytes, and thymic stromal cells. CD54 is also present on endothelial cells and such non-immune cells as hepatocytes, cardiomyocytes, and vascular smooth muscle cells. In addition, reports demonstrate that CD54 expression can be induced in vitro by proinflammatory cytokines, including IFN-gamma, IL-1, CCL2, or TNF-alpha. CD54 plays important roles in transendothelial migration of leukocytes to sites of inflammation and in T cell interactions with antigen presenting cells. Studies indicate that CD54 mediates these functions by binding to lymphocyte function-associated molecule (LFA-1; also known as CD11a/CD18). The 1A29 antibody has been reported to block interactions between CD54 and LFA-1. Applications Reported: This 1A29 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1A29 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be ca...

ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD54 (ICAM-1)
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q00238
ICAM1
Affinity chromatography
RUO
25464
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
1A29
PerCP-eFluor 710
ICAM1
BB2; CD54; cell surface glycoprotein P3.58; Human rhinovirus receptor; ICAM; Icam1; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; intercellular adhesion molecule-1; intercellular adhesion molecule-1 precursor; Ly 47; Ly-47; major group rhinovirus receptor; MALA2; MALA-2; MyD10; P3.58; sCD54; soluble CD54; Surface antigen of activated B cells
Mouse
25 μg
Primary
Rat
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.