missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD53 Antibody (MEM-53), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-393AF350
This item is not returnable.
View return policy
Description
CD53 Monoclonal specifically detects CD53 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| CD53 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CD53 | |
| Leukocytes of patient suffering from a LGL-type leukemia. | |
| Primary | |
| The antibody MEM-53 reacts with CD53, a 32-40 kDa tetraspanin family glycoprotein exclusivelly expressed on leukocytes; it is not present on platelets, red blood cells and non-hematopoietic cells. The antibody MEM-53 reacts also with deglycosylated molecule (molecular weight of the antigen is reduced by 15 kDa using endoglycosidase F). HLDA IV; WS Code NL 59 HLDA V; WS Code B CD53.5 HLDA V; WS Code BP BP287 HLDA V; WS Code T T-096 HLDA V; WS Code X XB004 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| MEM-53 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53 | |
| Mouse | |
| 0.1 mL | |
| 963 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction