missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD52 Rabbit anti-Human, Monkey, Clone: CD52/2276R, Novus Biologicals™
Rabbit Monoclonal Antibody
£431.00
Specifications
| Antigen | CD52 |
|---|---|
| Clone | CD52/2276R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
CD52 Monoclonal specifically detects CD52 in Human, Monkey samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| CD52 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Monkey | |
| Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5 | |
| Recombinant full-length human CD52 protein | |
| Primary | |
| Store at 4°C. |
| CD52/2276R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 1043 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title