missing translation for 'onlineSavingsMsg'
Learn More

CD52 Antibody (YTH 34.5-G2b (Campath-1G)), DyLight 350, Novus Biologicals™

Product Code. 18350691 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350691 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350691 Supplier Novus Biologicals Supplier No. NBP275899UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CD52 Monoclonal specifically detects CD52 in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD52
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone YTH 34.5-G2b (Campath-1G)
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5
Gene Symbols CD52
Host Species Rat
Immunogen Human peripheral mononuclear cells enriched for T-cells.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1043.0
Test Specificity Human CD52 antigen, also known as CAMPATH-1. CD52 is expressed at high density by lymphocytes, monocytes, eosinophils, thymocytes and macrophages. It is expressed by most lymphoid derived malignancies, although expression on myeloma cells is variable.
Target Species Human, Cynomolgus Monkey, Rhesus Macaque
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.