missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD52, anti-Human, Clone: Campath-1H, Abnova™
Monoclonal Antibody
Brand: Abnova RAB00044.200ug
This item is not returnable.
View return policy
Specifications
| CD52 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (0.02% Proclin 300). | |
| CDW52 | |
| Original antibody is raised against human lymphocytes. | |
| RUO | |
| 1043 | |
| Store at 4°C for up to 3 months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Flow Cytometry | |
| Campath-1H | |
| Flow Cytometry (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CD52 molecule | |
| CD52 | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction