missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD50 (ICAM-3) Polyclonal Antibody

Rabbit Polyclonal Antibody
£133.00 - £397.00
Specifications
| Antigen | CD50 (ICAM-3) |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17720624
![]() |
Invitrogen™
PA5119214 |
200 μL |
£397.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17773504
|
Invitrogen™
PA5122109 |
20 μL |
£133.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD50 (ICAM-3) Polyclonal Antibody for Western Blot, IHC (P)
CD50 (intracellular adhesion molecule 3, ICAM-3) is a transmembrane glycoprotein expressed by leukocytes, that serves as a counter-receptor for the lymphocyte function-associated antigen (LFA)-1 integrin. Besides functioning as an adhesive molecule that mediates e.g. the contact between T cells and antigen presenting cells, ICAM-3 regulates affinity of LFA-1 for ICAM-1 and induces T cell activation and proliferation. ICAM-3 plays an essential role in the initiation of the immune response both on T cells and antigen presenting cells and interacts also with CD209 (dendritic cell-specific ICAM-3-grabbing nonintegrin, DC-SIGN), a C-type lectin of dendritic cells and macrophages; this process is involved in dialogue between dendritic cells and granulocytes.Specifications
| CD50 (ICAM-3) | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| ICAM3 | |
| CD50; CDw50; ICAM3; ICAM-3; ICAM-R; Intercellular adhesion molecule 3; intercellular adhesion molecule-3 | |
| ICAM3 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P32942 | |
| 3385 | |
| Recombinant protein Intercellular Adhesion Molecule 3. The antigen corresponds to amino acid range 46-197 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title