missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Rat, FITC, Clone: 53-7.3, enQuireBio™
Rat Monoclonal Antibody
£62.45 - £164.00
Specifications
Antigen | CD5 |
---|---|
Clone | 53-7.3 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961528
|
EnquireBio
QAB13-F-25UG |
25 μg |
£62.45
25µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951528
|
EnquireBio
QAB13-F-100UG |
100 μg |
£164.00
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD5 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
12507 | |
IgG2a κ | |
Protein A/G Purified |
53-7.3 | |
Flow Cytometry | |
FITC | |
RUO | |
XP_006526684 | |
Cd5 | |
Primary | |
Store at 2-4°C. |