missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Mouse, anti-Human, FITC, Clone: UCHT2, STEMCELL Technologies
Mouse Monoclonal Antibody
£144.45 - £322.65
Specifications
| Antigen | CD5 |
|---|---|
| Clone | UCHT2 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17118931
|
Stemcell Technologies
60082FI.1 |
25 Tests |
£144.45
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17108931
|
Stemcell Technologies
60082FI |
100 Tests |
£322.65
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The UCHT2 antibody reacts with CD5, an ∼67 kDa single-chain type I glycoprotein and member of the scavenger receptor superfamily, which is constitutively expressed on thymocytes, T cells, B cell subsets, peripheral blood dendritic cells and some leukemia and lymphoma cells, including chronic B lymphocytic leukemia (B-CLL) cells
- CD5 is expressed at low levels on thymocytes and at high density on mature T cells
- Putative ligands include CD5L and CD72
- CD5 modulates T and B cell receptor signaling, thymocyte maturation, and T cell/B cell interactions via its physical and functional associations with the T cell receptor/CD3 complex and the B cell receptor
- Studies with CD5 knockout mice indicate that CD5 negatively regulates antigen receptor-mediated signaling in thymocytes and mature T cells
- The UCHT2 antibody binds to epitope 2 located in the extracellular domain of CD5
Specifications
| CD5 | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry | |
| FITC | |
| Cynomolgus, Rhesus Monkey, Chimpanzee, Human | |
| Primary |
| UCHT2 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title