Learn More
CD5 Monoclonal Antibody (53-7.3), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£72.50 - £145.00
Specifications
Antigen | CD5 |
---|---|
Clone | 53-7.3 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15226767
|
Affymetrix eBioscience
14-0051-82 |
100 μg |
£72.50
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15236767
|
Affymetrix eBioscience
14-0051-85 |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The 53-7.3 monoclonal antibody reacts with mouse CD5, a 67 kDa protein expressed by a majority of thymocytes, mature T cells and a subset of B cells. The expression of CD5 by a small subset of B cells characterizes a developmentally and functionally distinct lineage of B cells called B-1 cells. CD5 is a counter-receptor for CD72 and plays a role in the T-B cell interaction. Applications Reported: The 53-7.3 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining. Applications Tested: The 53-7.3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter's Syndrome.Specifications
CD5 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1 | |
Cd5 | |
Primary | |
4° C | |
Cd5 |
53-7.3 | |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rat | |
Mouse | |
P13379 | |
12507 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.