missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Antibody (C5/473 + CD5/54/F6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34583-0.1mg
This item is not returnable.
View return policy
Description
CD5 Monoclonal antibody specifically detects CD5 in Human, Rat, Bovine, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CD5 | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1-2 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
| Human CD5 recombinant protein (C5/473); A synthetic peptide from the intracellular region of human CD5 (CD5/54/F6) | |
| 0.1 mg | |
| Adaptive Immunity, Cell Biology, Cytokine Research, Immunology | |
| 921 | |
| Store at -20°C to -80°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| C5/473 + CD5/54/F6 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Bovine, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction