missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Antibody (7.8), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14758PE
This item is not returnable.
View return policy
Description
CD5 Monoclonal antibody specifically detects CD5 in Human, Mouse (Negative) samples. It is validated for Flow Cytometry, Flow (Cell Surface)
Specifications
| CD5 | |
| Monoclonal | |
| PE | |
| CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
| Human activated PBMC | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology | |
| 921 | |
| Store at 4°C in the dark. | |
| IgG2 α |
| Flow Cytometry | |
| 7.8 | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction