missing translation for 'onlineSavingsMsg'
Learn More

CD49f (Integrin α6) Rat, anti-Mouse, Pacific Blue, Clone: GoH3, STEMCELL Technologies

Rat Monoclonal Antibody

£252.45 - £523.80

Specifications

Antigen CD49f (Integrin α6)
Clone GoH3
Applications Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay
Classification Monoclonal
Conjugate Pacific Blue
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17118701
Encompass_Preferred
View Documents
Stemcell Technologies
60037PB.1
25 Tests
£252.45
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17108701
Encompass_Preferred
View Documents
Stemcell Technologies
60037PB
100 Tests
£523.80
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The GoH3 antibody reacts with CD49f (integrin α6), an ∼150 kDa transmembrane glycoprotein that associates non-covalently with CD29 (integrin β1) or CD104 (integrin β4) to form the heterodimeric receptors VLA-6 and α6β4, which bind the extracellular matrix protein laminin
  • CD49f is a disulfide-linked dimer comprising an ∼120 kDa heavy chain and an ∼30 kDa membrane-bound light chain
  • Splice variants exist, which affect the cytoplasmic domain of the protein
  • CD49f is expressed on the surface of T cells, monocytes, platelets, placental trophoblasts, epithelial cells, and endothelial cells
  • It is involved in cell adhesion and regulating signaling pathways involved in a variety of processes, including the activation and proliferation of T cells, and the differentiation and maintenance of stem cell pluripotency
  • CD49f is considered the most important marker for selecting mouse mammary stem and progenitor cells
  • The GoH3 antibody reacts with an extracellular epitope on CD49f
Specifications

Specifications

CD49f (Integrin α6)
Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay
Pacific Blue
Baboon, Cynomolgus, Pig, Human
Primary
GoH3
Monoclonal
Rat
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.