missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD49f (Integrin α6) Rat, anti-Mouse, APC, Clone: GoH3, STEMCELL Technologies
Rat Monoclonal Antibody
POA - £535.95
Specifications
| Antigen | CD49f (Integrin α6) |
|---|---|
| Clone | GoH3 |
| Applications | Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17158691
|
Stemcell Technologies
60037AZ.1 |
25 Tests | |||||||
17148691
|
Stemcell Technologies
60037AZ |
100 Tests |
£535.95
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The GoH3 antibody reacts with CD49f (integrin α6), an ∼150 kDa transmembrane glycoprotein that associates non-covalently with CD29 (integrin β1) or CD104 (integrin β4) to form the heterodimeric receptors VLA-6 and α6β4, which bind the extracellular matrix protein laminin
- CD49f is a disulfide-linked dimer comprising an ∼120 kDa heavy chain and an ∼30 kDa membrane-bound light chain
- Splice variants exist, which affect the cytoplasmic domain of the protein
- CD49f is expressed on the surface of T cells, monocytes, platelets, placental trophoblasts, epithelial cells, and endothelial cells
- It is involved in cell adhesion and regulating signaling pathways involved in a variety of processes, including the activation and proliferation of T cells, and the differentiation and maintenance of stem cell pluripotency
- CD49f is considered the most important marker for selecting mouse mammary stem and progenitor cells
- The GoH3 antibody reacts with an extracellular epitope on CD49f
Specifications
| CD49f (Integrin α6) | |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| APC | |
| Baboon, Cynomolgus, Pig, Human | |
| Primary |
| GoH3 | |
| Monoclonal | |
| Rat | |
| IgG2a κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title