Learn More
Invitrogen™ CD49e (Integrin alpha 5) Recombinant Rabbit Monoclonal Antibody (0S9S6)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555213
Description
Immunogen Sequence: FNLDAEAPAV LSGPPGSFFG FSVEFYRPGT DGVSVLVGAP KANTSQPGVL QGGAVYLCPW GASPTQCTPI EFDSKGSRLL ESSLSSSEGE EPVEYKSLQW FGATVRAHGS SILACAPLYS WRTEKEPLSD PVGTCYLSTD NFTRILEYAP CRSDFSWAAG QGYCQGGFSA EFTKTGRVVL GGPGSYFWQG QILSATQEQI AESYYPEYLI NLVQGQLQTR QASSIYDDSY LGYSVAVGEF SGDDTEDFVA GVPKGNLTYG YVTILNGSDI RSLYNFSGEQ MASYFGYAVA ATDVNGDGLD DLLVGAPLLM DRTPDGRPQE VGRVYVYLQH PAGIEPTPTL TLTGHDEFGR FGSSLTPLGD LDQDGYNDVA IGAPFGGETQ QGVVFVFPGG PGGLGSKPSQ VLQPLWAASH TPDFFGSALR GGRDLDGNGY PDLIVGSFGV DKAVVYRGRP IVSASASLTI FPAMFNPEER SCSLEGNPVA CINLSFCLNA SGKHVADSIG FTVELQLDWQ KQKGGVRRAL FLASRQATLT QTLLIQNGAR EDCREMKIYL RNESEFRDKL SPIHIALNFS LDPQAPVDSH GLRPALHYQS KSRIEDKAQI LLDCGEDNIC VPDLQLEVFG EQNHVYLGDK NALNLTFHAQ NVGEGGAYEA ELRVTAPPEA EYSGLVRHPG NFSSLSCDYF AVNQSRLLVC DLGNPMKAGA SLWGGLRFTV PHLRDTKKTI QFDFQILSKN LNNSQSDVVS FRLSVEAQAQ VTLNGVSKPE AVLFPVSDWH PRDQPQKEED LGPAVHHVYE LINQGPSSIS QGVLELSCPQ ALEGQQLLYV TRVTGLNCTT NHPINPKGLE LDPEGSLHHQ QKREAPSRSS ASSGPQILKC PEAECFRLRC ELGPLHQQES QSLQLHFRVW AKTFLQREHQ PFSLQCEAVY KALKMPYRIL PRQLPQKERQ VATAVQWTKA EGSY.
The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
Specifications
| CD49e (Integrin alpha 5) | |
| Recombinant Monoclonal | |
| 0.29 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P08648 | |
| Itga5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 42-995 of human Integrin alpha 5 (ITGA5/CD49e). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 0S9S6 | |
| Unconjugated | |
| Itga5 | |
| 2-phospho-D-glycerate hydro-lyase; alpha enolase like 1; Alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; CD49 antigen-like family member E; Cd49e; C-myc promoter-binding protein; ENO1; ENO1L1; Enolase 1; enolase 1, (alpha); enolase-alpha; epididymis secretory protein Li 17; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; FNRA; HEL-S-17; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin subunit alpha 5; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); ITGA5; Itga5_retired; MBP-1; MBPB1; MPB1; MPB-1; MYC promoter-binding protein 1; NNE; Non-neural enolase; phosphopyruvate hydratase; Plasminogen-binding protein; PPH; tau-crystallin; very late activation protein 5, alpha subunit; VLA5; VLA-5; VLA5A | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3678 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.