missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD49e (Integrin α5 chain) Mouse anti-Human, BUV615, Clone: IIA1, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 752325
This item is not returnable.
View return policy
Description
CD49e (Integrin α5 chain) Monoclonal antibody specifically detects CD49e (Integrin α5 chain) in Human samples. It is validated for Flow Cytometry.
Specifications
| CD49e (Integrin α5 chain) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Hu CD49e BUV615 IIA1 50 μg | |
| Human α5β1 Protein | |
| RUO | |
| CD49e (Integrin α5 chain) | |
| Refrigerate (2 to 8°C) | |
| Human |
| Flow Cytometry | |
| IIA1 | |
| Brilliant Ultraviolet 615 | |
| Mouse | |
| 1 x 50 μg | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction