missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD49d (Integrin alpha 4) Monoclonal Antibody (MRa4-1), APC, eBioscience™
GREENER_CHOICE

Mouse Monoclonal Antibody

£223.00

Specifications

Antigen CD49d (Integrin alpha 4)
Clone MRa4-1
Concentration 0.2 mg/mL
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Applications Flow Cytometry
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15558016
GREENER_CHOICE
View Documents
Invitrogen™
17-1490-82
100 μg
£223.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The MRa4-1 monoclonal antibody reacts with CD49d, which is also known as the integrin alpha 4 subunit. This 150-kDa cell surface protein is expressed by thymocytes, peripheral lymphocytes, monocytes, and eosinophils in complex with either integrin beta1 or integrin beta7. Outside of the immune system, CD49d is also expressed on mast cells, neural crest cells, retinal cells, and dorsal root ganglion neurons. Integrins are responsible for lymphocyte homing to Peyer's patches and mesenteric lymph nodes, as well as neural development and regeneration. Several ligands have been reported for the integrin a4b1 complex, including VCAM-1 (CD106) and fibronectin. The MRa1-4 antibody has been reported to inhibit fibronectin binding and lymphocyte migration. Applications Reported: This MRa4-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MRa4-1 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

The integrin alpha 4 (also known as CD49d and ITGA4) belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha and beta chains. Alpha4 chain associates with either beta1 or beta7 chain. It has been demonstrated that the putative ligand-binding sites of both integrin alpha4beta1 and alpha4beta7 is located on the alpha4 chain. These ligands included Madcam, VCAM, and fibronectin. Madcam is known as the principal ligand for integrin alpha4beta7. Recently it was also demonstrated that HIV-1 envelope can mimic Madcam by binding to and signaling though integrin alpha4beta7, the gut mucosal homing receptor for peripheral T cells.
Specifications

Specifications

CD49d (Integrin alpha 4)
0.2 mg/mL
Flow Cytometry
APC
Mouse
RUO
PBS with 0.09% sodium azide; pH 7.2
0
311144
Primary
Affinity chromatography
MRa4-1
4°C, store in dark, DO NOT FREEZE!
Monoclonal
Liquid
IgG2a κ
Rat
ITGA4
269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha
ITGA4
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.