Learn More
CD49b (Integrin alpha 2) Monoclonal Antibody (HMa2), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 16-0491-025
Description
The HMalpha2 monoclonal antibody reacts with mouse CD49b, the 150 kDa integrin alpha2 subunit. The complex of CD49b non-covalently associated with integrin beta1 (CD29), also known as VLA-2, is a receptor for collagen and laminin. This complex is expressed by some CD4^+ T cells, IEL, NK cells, platelets and epithelial cells. Applications Reported: The HM alpha2 antibody has been reported for use in flow cytometric analysis. It has also been reported for use in in vitro functional assays. Applications Tested: The HM alpha2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
CD49b (Integrin alpha 2) Monoclonal antibody specifically detects CD49b (Integrin alpha 2) in Mouse samples. It is validated for Flow Cytometry, Functional Assay
Specifications
| CD49b (Integrin alpha 2) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| Q62469 | |
| Armenian Hamster | |
| 25 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Functional Assay | |
| HMa2 | |
| Functional Grade | |
| Itga2 | |
| alpha 2 subunit of VLA-2 receptor; BDPLT9; BR; CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; HPA-5; human platelet alloantigen system 5; integrin alpha 2; integrin alpha-2; integrin subunit alpha 2; integrin, alpha 2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); ITGA2; platelet antigen Br; platelet glycoprotein GPIa; platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2; VLA2 receptor; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha; VLAA2 | |
| Affinity chromatography | |
| RUO | |
| 16398 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.