missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD49b (Integrin alpha 2) Monoclonal Antibody (DX5), PE-Cyanine5.5, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£169.00 - £419.00
Specifications
| Antigen | CD49b (Integrin alpha 2) |
|---|---|
| Clone | DX5 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16676334
|
Invitrogen™
35-5971-80 |
25 μg |
£169.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16686334
![]() |
Invitrogen™
35-5971-82 |
100 μg |
£419.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD49b (Integrin alpha 2, Integrin alpha 2 chain, VLA-2 alpha chain, HM alpha 2) is a member of the integrin family. It is a glycoprotein with molecular weight of 150 kD, and it complexes with CD29 (Integrin beta 1) to form the heterodimeric integrin VLA-2 (integrin alpha 2 beta 1, or GPIa/IIa) complex. VLA-2 is an extracellular receptor for laminin, collagen, and fibronectin, and interaction with its ligands results in the activation of intracellular signaling pathways. It has reported roles in VEGF-induced angiogenesis in vivo, as well as adhesion and lymphocyte activation. CD49b is expressed by NK cells, NK-T cells, monocytes, platelets, and epithelial cells. It is also expressed on adaptive immune cells such as T and B cells, specifically on a subset of CD4+ T cells in the spleen, on intraepithelial and lamina propria lymphocytes in the intestine, as well as on a population of peripheral CD4+ type 1 T regulatory (Tr1) cells that co-express LAG-3.Specifications
| CD49b (Integrin alpha 2) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| alpha 2 subunit of VLA-2 receptor; BDPLT9; BR; CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; HPA-5; human platelet alloantigen system 5; integrin alpha 2; integrin alpha-2; integrin subunit alpha 2; integrin, alpha 2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); Itga2; platelet antigen Br; platelet glycoprotein GPIa; platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2; VLA2 receptor; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha; VLAA2 | |
| Itga2 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Itga2 |
| DX5 | |
| Flow Cytometry | |
| PE-Cyanine5.5 | |
| Rat | |
| Mouse | |
| Q62469 | |
| 16398 | |
| IgM κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title