Learn More
CD49b (Integrin alpha 2) Monoclonal Antibody (HMa2), eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen 14-0491-85
Description
Description: The HMalpha2 monoclonal antibody reacts with mouse CD49b, the 150 kDa integrin alpha2 subunit. The complex of CD49b non-covalently associated with integrin beta1 (CD29), also known as VLA-2, is a receptor for collagen and laminin. This complex is expressed by some CD4^+ T cells, IEL, NK cells, platelets and epithelial cells. Applications Reported: The HM alpha2 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining of frozen tissue (IHC-F). It has also been reported for use in in vitro functional assays (Please use Functional Grade purified HMa2, cat. 16-0491, in functional assays.). Applications Tested: The HM alpha2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
GTP-binding proteins (G-proteins) play a critical role in signal transduction by coupling receptors to effectors. The intracellular consequences of G-protein activation include second messenger generation, protein phosphorylation, ion channel activation, gene induction, cell growth, and differentiation. Gi belongs to the pertussis toxin sensitive G-proteins. Gi consists of three different subtypes of a-subunits (ai1, ai2 and ai3).
Specifications
| CD49b (Integrin alpha 2) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q62469 | |
| Itga2 | |
| Affinity chromatography | |
| RUO | |
| 16398 | |
| 4° C | |
| Liquid |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation | |
| HMa2 | |
| Unconjugated | |
| Itga2 | |
| BR, CD49B, GPIa, VLA-2, VLAA2, ITGA2 | |
| Armenian Hamster | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.