missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD48 (SLAMF2) Hamster, anti-Mouse, APC, Clone: HM48-1, STEMCELL Technologies
Hamster Monoclonal Antibody
£160.65 - £500.85
Specifications
| Antigen | CD48 (SLAMF2) |
|---|---|
| Clone | HM48-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17159221
|
Stemcell Technologies
60162AZ.1 |
25 μg |
£160.65
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17149221
|
Stemcell Technologies
60162AZ |
100 μg |
£500.85
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The HM48-1 antibody reacts with mouse CD48, a 45 kDa GPI-anchored glycoprotein and a member of the signaling lymphocyte activation molecule (SLAM) family and Ig superfamily
- SLAM family members play a role in multiple aspects of immune function including costimulation, cytotoxicity, and cytokine production
- CD48 also plays a role in cell adhesion and T cell activation
- In mice, CD48 is thought to be present on nearly all hematopoietic cells except neutrophils and ESLAM cells (CD45+EPCR+CD48-CD150+ hematopoietic stem cells with long-term self-renewal capacity)
- Ligands for CD48 include CD2 and CD244, binding of which helps to promote interactions between immune cells
- HM48-1 is reported to be capable of blocking CD48:CD2 and CD48:CD244 interactions in vitro and in vivo
- HM48-1 is also reported to inhibit the proliferative response of spleen cells to mitogens, inhibit CTL activity, and enhance the survival of cardiac allografts
Specifications
| CD48 (SLAMF2) | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Primary |
| HM48-1 | |
| Monoclonal | |
| Hamster | |
| IgG1 λ3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title