missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD48/SLAMF2 Antibody (Regeneron patent anti-CD48) - IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28815-100ug
This item is not returnable.
View return policy
Description
CD48/SLAMF2 Monoclonal antibody specifically detects CD48/SLAMF2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD48/SLAMF2 | |
| Monoclonal | |
| Unconjugated | |
| P09326 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| Regeneron patent anti-CD48 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2 | |
| CD48 | |
| 100 μg | |
| Cell Biology, Cellular Markers, Immunology, Stem Cell Markers | |
| 962 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction