missing translation for 'onlineSavingsMsg'
Learn More

CD48/SLAMF2 Antibody (341), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18379740 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379740 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379740 Supplier Novus Biologicals Supplier No. NBP290577AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD48/SLAMF2 Monoclonal specifically detects CD48/SLAMF2 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Sandwich ELISA Detection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD48/SLAMF2
Applications Flow Cytometry, Immunofluorescence, ELISA
Classification Monoclonal
Clone 341
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD48/SLAMF2 (NP_031675.1; Met1-Arg216).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 962
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.