missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD48 Recombinant Rabbit Monoclonal Antibody (248), APC

Rabbit Recombinant Monoclonal Antibody
£107.00 - £332.00
Specifications
| Antigen | CD48 |
|---|---|
| Clone | 248 |
| Concentration | 0.1 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30535120
![]() |
Invitrogen™
MA560294 |
100 Tests |
£332.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30534861
|
Invitrogen™
MA560295 |
25 Tests |
£107.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD48 Recombinant Monoclonal Antibody for Flow
CD48, also known as Blast-1, is a 45 kDa glycoprotein that is part of the SLAM and Ig superfamilies. It is a GPI-anchored protein broadly expressed on most hematopoietic cells, including lymphocytes and monocytes, but is weakly expressed on granulocytes and absent on platelets, fibroblasts, epithelium, and endothelium. CD48 plays a critical role in adhesion and T cell activation, serving as a high-affinity ligand for 2B4 and a low-affinity ligand for CD2. The interaction between 2B4 and CD48 among NK and NK-T cells regulates cell proliferation. CD48 is involved in eosinophil activation and its expression increases in several infectious diseases. It is also a marker for detecting defects in the GPI anchoring structure in patients with paroxysmal nocturnal hemoglobulinuria (PNH). Recent studies have highlighted the differential expression of SLAM family members, including CD48, CD150, and CD244, among bone marrow hematopoietic progenitors. This provides a useful tool for predicting the primitiveness of hematopoietic progenitors: hematopoietic stem cells (HSCs) are CD150(+)CD244(-)CD48(-), non-self-renewing multipotent progenitors (MPPs) are CD244(+)CD150(-)CD48(-), and the most restricted progenitors are CD48(+)CD244(+)CD150(-). Additionally, the 2B4-CD48 interaction may be involved in the pathology of allergic airway diseases, offering a potential pharmacological target for treating conditions like allergic rhinitis and asthma.Specifications
| CD48 | |
| 0.1 mg/mL | |
| Flow Cytometry | |
| APC | |
| Rabbit | |
| RUO | |
| PBS with 0.5% BSA and 0.09% sodium azide | |
| P09326 | |
| 962 | |
| Recombinant Human CD48/SLAMF2 protein (Met1-Ser220). | |
| Antibody |
| 248 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Cd48 | |
| AI449234; AW610730; B-cell activation marker; BCM1; Bcm-1; BCM1 surface antigen; BLAST; BLAST1; BLAST-1; B-lymphocyte activation marker BLAST-1; Cd48; CD48 antigen; CD48 antigen (B cell membrane protein); CD48 antigen (B-cell membrane protein); Cd48 molecule; CD48d; hCD48; HM48-1; leukocyte antigen MEM-102; mCD48; MEM-102; MRC OX-45 surface antigen; sCD 48; sCD48; Sgp-60; Signaling lymphocytic activation molecule 2; Signaling lymphocytic activation molecule 2 (SLAMF2); SLAM family member 2; SLAMF2; soluble CD 48; soluble CD48; TCT.1 | |
| Cd48 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title